Thermo Scientific Chemicals

Maleic Acid, 99%, Thermo Scientific Chemicals

Catalog number: 125230051
5 kg, Plastic drum, Each
Thermo Scientific Chemicals

Maleic Acid, 99%, Thermo Scientific Chemicals

Catalog number: 125230051
5 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 125230051
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-16-7
IUPAC Name
(2E)-but-2-enedioic acid
Molecular Formula
C4H4O4
InChI Key
VZCYOOQTPOCHFL-OWOJBTEDSA-N
SMILES
OC(=O)\C=C\C(O)=O
Appearance (Form)
Crystalline powder or granules and/or chunks
Titration with NaOH
98.5 to 101.5 % (on dry substance)
HPLC
>=98.5 %
Infrared spectrum
Conforms
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text