Thermo Scientific Chemicals

Maleic anhydride, 99%, pastilles, Thermo Scientific Chemicals

Catalog number: 125240250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Maleic anhydride, 99%, pastilles, Thermo Scientific Chemicals

Catalog number: 125240250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 125240250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-31-6
IUPAC Name
2,5-dihydrofuran-2,5-dione
Molecular Formula
C4H2O3
InChI Key
FPYJFEHAWHCUMM-UHFFFAOYSA-N
SMILES
O=C1OC(=O)C=C1
Infrared spectrum
Conforms
Appearance (Form)
Pastilles
Melting point
52°C to 55°C
Free acid
=<0.5 % (as maleic acid)
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text