DL-Malic acid, 99+%
DL-Malic acid, 99+%
DL-Malic acid, 99+%
Thermo Scientific Chemicals

DL-Malic acid, 99+%

CAS: 6915-15-7 | C4H6O5 | 134.087 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 125252500
Price (USD)
40.65
Online Exclusive
45.00
Save 4.35 (10%)
Each
In stock
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
40.65
Online Exclusive
45.00
Save 4.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS6915-15-7
IUPAC Name2-hydroxybutanedioic acid
Molecular FormulaC4H6O5
InChI KeyBJEPYKJPYRNKOW-UHFFFAOYNA-N
SMILESOC(CC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Titration with NaOH>=99.0 %
Insoluble matter=<0.1 % (in water)
Impurity=<1.0 % Fumaric acid
Melting point128°C to 132°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link