Thermo Scientific Chemicals

Malonic acid, 99%, Thermo Scientific Chemicals

Catalog number: 125260010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Malonic acid, 99%, Thermo Scientific Chemicals

Catalog number: 125260010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 125260010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
141-82-2
IUPAC Name
propanedioic acid
Molecular Formula
C3H4O4
InChI Key
OFOBLEOULBTSOW-UHFFFAOYSA-N
SMILES
OC(=O)CC(O)=O
Appearance (Color)
White
Titration with NaOH
>=98.5 %
Melting point
133°C to 136°C
Sulfate (SO4)
=<500 ppm
Chloride (Cl)
=<500 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text