Malonic acid, 99%
Malonic acid, 99%
Malonic acid, 99%
Thermo Scientific Chemicals

Malonic acid, 99%

CAS: 141-82-2 | C3H4O4 | 104.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 125260051
Price (USD)
825.65
Special offer
Online exclusive
Ends: 26-Sep-2025
1,020.00
Save 194.35 (19%)
Each
Add to cart
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
825.65
Special offer
Online exclusive
Ends: 26-Sep-2025
1,020.00
Save 194.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS141-82-2
IUPAC Namepropanedioic acid
Molecular FormulaC3H4O4
InChI KeyOFOBLEOULBTSOW-UHFFFAOYSA-N
SMILESOC(=O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with NaOH>=98.5 %
Sulfate (SO4)=<500 ppm
Chloride (Cl)=<500 ppm
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link