Malonic acid, 99%, Thermo Scientific Chemicals
Malonic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Malonic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 125262500
Price (USD)/ Each
102.65
Online exclusive
120.00 
Save 17.35 (14%)
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
102.65
Online exclusive
120.00 
Save 17.35 (14%)
Add to cart
Malonic acid, 99%, Thermo Scientific Chemicals
Catalog number125262500
Price (USD)/ Each
102.65
Online exclusive
120.00 
Save 17.35 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Namepropanedioic acid
Molecular FormulaC3H4O4
InChI KeyOFOBLEOULBTSOW-UHFFFAOYSA-N
SMILESOC(=O)CC(O)=O
Molecular Weight (g/mol)104.06
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with NaOH>=98.5 %
Melting point133°C to 136°C
Sulfate (SO4)=<500 ppm
Chloride (Cl)=<500 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only