Thermo Scientific Chemicals

Malononitrile, 99%, Thermo Scientific Chemicals

Catalog number: 125270025
2.5 kg, Glass bottle, Each
Thermo Scientific Chemicals

Malononitrile, 99%, Thermo Scientific Chemicals

Catalog number: 125270025
2.5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 125270025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
109-77-3
IUPAC Name
propanedinitrile
Molecular Formula
C3H2N2
InChI Key
CUONGYYJJVDODC-UHFFFAOYSA-N
SMILES
N#CCC#N
Appearance (Form)
Low melting crystalline mass
Melting point
30°C to 34°C
Appearance (Color)
White to yellow-brown
Infrared spectrum
Conforms
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text