Thermo Scientific Chemicals

Malonyl dichloride, 97%, Thermo Scientific Chemicals

Catalog number: 125281000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Malonyl dichloride, 97%, Thermo Scientific Chemicals

Catalog number: 125281000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 125281000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1663-67-8
IUPAC Name
propanedioyl dichloride
Molecular Formula
C3H2Cl2O2
InChI Key
SXYFKXOFMCIXQW-UHFFFAOYSA-N
SMILES
ClC(=O)CC(Cl)=O
Appearance (Color)
Clear yellow to orange to brown
Infrared spectrum
Conforms
Refractive index
1.4620 to 1.4660 (20°C, 589 nm)
Appearance (Form)
Liquid
Titration Argentometric
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text