(S)-(+)-Mandelic Acid ≥99%, Thermo Scientific Chemicals
(S)-(+)-Mandelic Acid ≥99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(S)-(+)-Mandelic Acid ≥99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number 125301000
Price (USD)/ Each
72.65
Online exclusive
84.90 
Save 12.25 (14%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
72.65
Online exclusive
84.90 
Save 12.25 (14%)
Add to cart
(S)-(+)-Mandelic Acid ≥99%, Thermo Scientific Chemicals
Catalog number125301000
Price (USD)/ Each
72.65
Online exclusive
84.90 
Save 12.25 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Name(2S)-2-hydroxy-2-phenylacetic acid
Molecular FormulaC8H8O3
InChI KeyIWYDHOAUDWTVEP-ZETCQYMHSA-N
SMILESO[C@H](C(O)=O)C1=CC=CC=C1
Molecular Weight (g/mol)152.15
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Titration with NaOH>=99.0 %
Melting point130°C to 135°C
Appearance (Form)Powder or crystals or flakes
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only