DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

DL-Mandelic acid, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
250 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number 125310010
Price (USD)/ Each
118.65
Online exclusive
132.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
118.65
Online exclusive
132.00 
Save 13.35 (10%)
Add to cart
DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
Catalog number125310010
Price (USD)/ Each
118.65
Online exclusive
132.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS90-64-2
IUPAC Name2-hydroxy-2-phenylacetic acid
Molecular FormulaC8H8O3
InChI KeyIWYDHOAUDWTVEP-UHFFFAOYNA-N
SMILESOC(C(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
Titration with NaOH>=99.0 %
Appearance (Form)Crystalline powder or crystals
Melting point117°C to 121°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only