1x1 image pixel for data collection
Cholesterol Lipid Concentrate (250X)
Product Image
Gibco™

Cholesterol Lipid Concentrate (250X)

250X Cholesterol Lipid Concentrate is a protein-free, chemically-defined supplement used for growing NSO and other lipid dependent cell lines toRead more
Have Questions?
Catalog NumberQuantity
12531018100 mL
Catalog number 12531018
Price (USD)
286.00
Each
In stock
Add to cart
Quantity:
100 mL
Price (USD)
286.00
Each
Add to cart
250X Cholesterol Lipid Concentrate is a protein-free, chemically-defined supplement used for growing NSO and other lipid dependent cell lines to high densities while supporting growth and protein expression without complicating downstream purification. The supplement can be added to 1X media such as CD Hybridoma and filtered with minimal loss of lipids. Custom formulations and packaging configurations as well as supplements used in feeding strategies are available through our Custom Products Services group.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeMammalian Cells
Concentration250 X
Culture TypeMammalian Cell Culture, Insect Cell Culture
Quantity100 mL
Shelf Life18 Months
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
FormLiquid
Product TypeCholesterol Lipid Concentrate
SterilitySterile
pH6 to 8
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link