Thermo Scientific Chemicals

(R)-(-)-Mandelic Acid 99%, Thermo Scientific Chemicals

Catalog number: 125321000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

(R)-(-)-Mandelic Acid 99%, Thermo Scientific Chemicals

Catalog number: 125321000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 125321000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
611-71-2
IUPAC Name
(2R)-2-hydroxy-2-phenylacetic acid
Molecular Formula
C8H8O3
InChI Key
IWYDHOAUDWTVEP-SSDOTTSWSA-N
SMILES
O[C@@H](C(O)=O)C1=CC=CC=C1
Appearance (Color)
White to light yellow to light beige
Melting point
130°C to 135°C
Water
=<1 % (K.F.)
Specific optical rotation
-152° to -158° (20°C, 589 nm) (c=2, H2O)
Appearance (Form)
Crystalline powder or crystals or flakes

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text