D-Mannitol, 98+%
Thermo Scientific Chemicals

D-Mannitol, 98+%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
100 g
500 g
1 kg
2.5 kg
Packaging:
View all
Plastic drum
Catalog number 125340050
Price (USD)/ Each
255.65
Online exclusive
284.00 
Save 28.35 (10%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
255.65
Online exclusive
284.00 
Save 28.35 (10%)
Add to cart
D-Mannitol, 98+%
Catalog number125340050
Price (USD)/ Each
255.65
Online exclusive
284.00 
Save 28.35 (10%)
-
Add to cart
Chemical Identifiers
CAS69-65-8
IUPAC Namehexane-1,2,3,4,5,6-hexol
Molecular FormulaC6H14O6
InChI KeyFBPFZTCFMRRESA-UHFFFAOYNA-N
SMILESOCC(O)C(O)C(O)C(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Nickel (Ni)=<1 ppm
Appearance (Color)White
Melting point165°C to 170°C
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only