Thermo Scientific Chemicals

D(+)-Melibiose monohydrate, 99+%

Catalog number: 125370250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

D(+)-Melibiose monohydrate, 99+%

Catalog number: 125370250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 125370250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
66009-10-7
IUPAC Name
2,3,4,5-tetrahydroxy-6-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanal hydrate
Molecular Formula
C12H24O12
InChI Key
CHIDEFLSUMQFBY-UHFFFAOYNA-N
SMILES
O.OCC1OC(OCC(O)C(O)C(O)C(O)C=O)C(O)C(O)C1O
Appearance (Color)
White to almost white
HPLC
>=99.0 %
Loss on drying
3.5 % to 6.0 % (1 g, 105°C)
Sulfated ash
=<0.1 %
Acidity
=<0.4 ml/6g (ml 0.1N NaOH/6g)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text