Chemical Identifiers
CAS66009-10-7
IUPAC Name2,3,4,5-tetrahydroxy-6-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanal hydrate
Molecular FormulaC12H24O12
InChI KeyCHIDEFLSUMQFBY-UHFFFAOYNA-N
SMILESO.OCC1OC(OCC(O)C(O)C(O)C(O)C=O)C(O)C(O)C1O
View more