Gateway® LR Clonase® enzyme mix contains a proprietary blend of Int (Integrase), IHF (Integration Host Factor) and Xis (Excisionase) enzymesRead more
Have Questions?
Change view
Catalog Number
Quantity
12538200
100 Reactions
12538120
20 Reactions
2 Options
Catalog number 12538200
Price (USD)/ 100 reactions
2,797.56
Special Offer
Online exclusive
Ends: 16-Dec-2024
3080.00
Save 282.44 (9%)
-
Add to cart
Quantity:
100 Reactions
Price (USD)/ 100 reactions
2,797.56
Special Offer
Online exclusive
Ends: 16-Dec-2024
3080.00
Save 282.44 (9%)
Add to cart
LR Clonase™ II Plus enzyme
Catalog number12538200
Price (USD)/ 100 reactions
2,797.56
Special Offer
Online exclusive
Ends: 16-Dec-2024
3080.00
Save 282.44 (9%)
-
Add to cart
Gateway® LR Clonase® enzyme mix contains a proprietary blend of Int (Integrase), IHF (Integration Host Factor) and Xis (Excisionase) enzymes that catalyze the in vitro recombination between an Entry clone (containing a gene of interest flanked by attL sites) and a Destination vector (containing attR sites) to generate your expression clone. We offer different formats of LR Clonase enzyme mixtures, depending on your application and desired format. LR Clonase II Plus enzyme mix is the latest version that offers the highest recombination efficiency available(Table 1), and is specifically optimized for both single and multiple fragment cloning (Table 1). LR Clonase II Plus enzyme mix is provided in an optimized single mix of enzyme and reaction buffer, ensuring enzyme stability and easy of use with few pippetting steps for both single and multiple fragment cloning. Our LR Clonase II enzyme mix is also available in a single mix format, while the original LR Clonase® enzyme is the enzyme and buffer in separate tubes
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Shipping ConditionDry Ice
EnzymeLR Clonase
Compatible BufferReaction Buffer
Quantity100 Reactions
Product TypeLR Clonase Enzyme
Product LineClonase™, Gateway™
Unit Size100 reactions
Contents & Storage
Includes LR Clonase II Plus enzyme and proteinase K solution (2 μg/μL). Store at -80°C. Guaranteed stable for 6 months when properly stored.