L(-)-Menthol, 99.5%
L(-)-Menthol, 99.5%
L(-)-Menthol, 99.5%
Thermo Scientific Chemicals

L(-)-Menthol, 99.5%

CAS: 2216-51-5 | C10H20O | 156.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
5 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 125400050
Price (USD)
23.50
Each
Estimated availability date  Pending  
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
23.50
Each
Add to cart
Chemical Identifiers
CAS2216-51-5
IUPAC Name(1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
Molecular FormulaC10H20O
InChI KeyNOOLISFMXDJSKH-KXUCPTDWSA-N
SMILESCC(C)[C@@H]1CC[C@@H](C)C[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=99.4 %
Specific optical rotation-48° to -51° (20°C, 589 nm) (c=10, EtOH)
Appearance (Form)Crystals or crystalline needles
Appearance (Color)Colorless to white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link