Thermo Scientific Chemicals

Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals

Catalog number: 125432500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals

Catalog number: 125432500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 125432500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
68-11-1
IUPAC Name
2-sulfanylacetic acid
Molecular Formula
C2H4O2S
InChI Key
CWERGRDVMFNCDR-UHFFFAOYSA-N
SMILES
OC(=O)CS
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Titration Iodimetric
97.5 to 102.5 %
Refractive index
1.5030 to 1.5050 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text