2-Mercaptobenzimidazole, 98%, Thermo Scientific Chemicals
2-Mercaptobenzimidazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Mercaptobenzimidazole, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 125441000
Price (USD)/ Each
22.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
26.30 
Save 3.65 (14%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
22.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
26.30 
Save 3.65 (14%)
Add to cart
2-Mercaptobenzimidazole, 98%, Thermo Scientific Chemicals
Catalog number125441000
Price (USD)/ Each
22.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
26.30 
Save 3.65 (14%)
-
Add to cart
Chemical Identifiers
CAS583-39-1
IUPAC Name2,3-dihydro-1H-1,3-benzodiazole-2-thione
Molecular FormulaC7H6N2S
InChI KeyYHMYGUUIMTVXNW-UHFFFAOYSA-N
SMILESS=C1NC2=CC=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Ash=<0.3 %
Volatile matter=<0.3 %
Insoluble matter=<0.3 % (in NaOH)
Appearance (Color)White to beige
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only