Thermo Scientific Chemicals

2-Mercaptobenzothiazole, 95%, Thermo Scientific Chemicals

Catalog number: 125452500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

2-Mercaptobenzothiazole, 95%, Thermo Scientific Chemicals

Catalog number: 125452500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 125452500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
149-30-4
IUPAC Name
2,3-dihydro-1,3-benzothiazole-2-thione
Molecular Formula
C7H5NS2
InChI Key
YXIWHUQXZSMYRE-UHFFFAOYSA-N
SMILES
S=C1NC2=CC=CC=C2S1
Appearance (Color)
Yellow
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
172°C to 180°C
Titration with NaOH
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text