2-Mercaptobenzoxazole, 99%, Thermo Scientific Chemicals
2-Mercaptobenzoxazole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Mercaptobenzoxazole, 99%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 125460250
Price (USD)/ Each
47.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
56.40 
Save 8.75 (16%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
47.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
56.40 
Save 8.75 (16%)
Add to cart
2-Mercaptobenzoxazole, 99%, Thermo Scientific Chemicals
Catalog number125460250
Price (USD)/ Each
47.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
56.40 
Save 8.75 (16%)
-
Add to cart
Chemical Identifiers
CAS2382-96-9
IUPAC Name2,3-dihydro-1,3-benzoxazole-2-thione
Molecular FormulaC7H5NOS
InChI KeyFLFWJIBUZQARMD-UHFFFAOYSA-N
SMILESS=C1NC2=CC=CC=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point192°C to 195°C
Titration Argentometric>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only