2-Mercaptoethanol, 99%, pure, Thermo Scientific Chemicals
2-Mercaptoethanol, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Mercaptoethanol, 99%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
10 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 125470025
Price (USD)/ Each
287.00
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
287.00
Add to cart
2-Mercaptoethanol, 99%, pure, Thermo Scientific Chemicals
Catalog number125470025
Price (USD)/ Each
287.00
-
Add to cart
Chemical Identifiers
CAS60-24-2
IUPAC Name2-sulfanylethan-1-ol
Molecular FormulaC2H6OS
InChI KeyDGVVWUTYPXICAM-UHFFFAOYSA-N
SMILESOCCS
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4990 to 1.5020 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Water=<0.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only