3-Mercapto-1,2-propanediol, 90 wt% aqueous solution, Thermo Scientific Chemicals
3-Mercapto-1,2-propanediol, 90 wt% aqueous solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Mercapto-1,2-propanediol, 90 wt% aqueous solution, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
500 mL
Packaging:
Glass bottle
Catalog number 125521000
Price (USD)/ Each
75.65
Online exclusive
89.00 
Save 13.35 (15%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
75.65
Online exclusive
89.00 
Save 13.35 (15%)
Add to cart
3-Mercapto-1,2-propanediol, 90 wt% aqueous solution, Thermo Scientific Chemicals
Catalog number125521000
Price (USD)/ Each
75.65
Online exclusive
89.00 
Save 13.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name3-sulfanylpropane-1,2-diol
Molecular FormulaC3H8O2S
InChI KeyPJUIMOJAAPLTRJ-UHFFFAOYNA-N
SMILESOCC(O)CS
Molecular Weight (g/mol)108.16
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Iodimetric>=88 %
Appearance (Form)Viscous solution
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only