3-Mercaptopropionic acid, 99+%, ACROS Organics™
3-Mercaptopropionic acid, 99+%, ACROS Organics™
Thermo Scientific Chemicals

3-Mercaptopropionic acid, 99+%, ACROS Organics™

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Catalog number 125531000
Price (USD)/ Each
-
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
IUPAC Name3-sulfanylpropanoic acid
Molecular FormulaC3H6O2S
InChI KeyDKIDEFUBRARXTE-UHFFFAOYSA-N
SMILESOC(=O)CCS
Molecular Weight (g/mol)106.14
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4910 to 1.4940 (20°C, 589 nm)
HPLC>=98.5 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only