Thermo Scientific Chemicals

DL-Mercaptosuccinic acid, 99%, Thermo Scientific Chemicals

Catalog number: 125551000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

DL-Mercaptosuccinic acid, 99%, Thermo Scientific Chemicals

Catalog number: 125551000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 125551000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
70-49-5
IUPAC Name
2-sulfanylbutanedioic acid
Molecular Formula
C4H6O4S
InChI Key
NJRXVEJTAYWCQJ-UHFFFAOYNA-N
SMILES
OC(=O)CC(S)C(O)=O
Melting point
150°C to 155°C
Appearance (Color)
White to almost white
Infrared spectrum
Conforms
Titration with NaOH
98.5 to 101.5 %
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text