DL-Mercaptosuccinic acid, 99%, Thermo Scientific Chemicals
DL-Mercaptosuccinic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

DL-Mercaptosuccinic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 125555000
Price (USD)/ Each
164.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
193.00 
Save 28.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
164.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
193.00 
Save 28.35 (15%)
Add to cart
DL-Mercaptosuccinic acid, 99%, Thermo Scientific Chemicals
Catalog number125555000
Price (USD)/ Each
164.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
193.00 
Save 28.35 (15%)
-
Add to cart
Chemical Identifiers
CAS70-49-5
IUPAC Name2-sulfanylbutanedioic acid
Molecular FormulaC4H6O4S
InChI KeyNJRXVEJTAYWCQJ-UHFFFAOYNA-N
SMILESOC(=O)CC(S)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point150°C to 155°C
Appearance (Color)White to almost white
Infrared spectrumConforms
Titration with NaOH98.5 to 101.5 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only