Thermo Scientific Chemicals

Mesitylene, 99%, Extra Pure, Thermo Scientific Chemicals

Catalog number: 125582500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

Mesitylene, 99%, Extra Pure, Thermo Scientific Chemicals

Catalog number: 125582500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 125582500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-67-8
IUPAC Name
1,3,5-trimethylbenzene
Molecular Formula
C9H12
InChI Key
AUHZEENZYGFFBQ-UHFFFAOYSA-N
SMILES
CC1=CC(C)=CC(C)=C1
Infrared spectrum
Conforms
Refractive index
1.4980 to 1.5000 (20°C, 589 nm)
Appearance (Color)
Clear colorless
GC
>=98.5 %
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text