VP-SFM AGT™ Medium
VP-SFM AGT™ Medium
Gibco™
VP-SFM AGT™ Medium
Gibco VP-SFM AGT Medium is a flexible-format medium that is equivalent to VP-SFM with the addition of Advanced Granulation Technology for simplified preparation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1255901910 L
125590271 L
1255900310 kg
12559001100 L
Catalog number 12559019
Price (USD)
896.00
Each
In stock
Add to cart
Quantity:
10 L
Customize this product
Price (USD)
896.00
Each
Add to cart
Gibco VP-SFM AGT Medium is a flexible-format medium that is equivalent to VP-SFM with the addition of Advanced Granulation Technology for simplified preparation. VP-SFM is a serum-free, animal-origin-free culture medium designed for growth of VERO cells and virus production. It is also suitable for the growth of COS-7, MDCK, BHK-21, and Hep2 cells and for recombinant protein production.
Features of VP-SFM AGT Medium include:
  • Serum-free, animal-origin-free, ultra-low protein formulation
  • Easy adaptation for many popular cell lines
  • Scalability in flasks, cell factories, and bioreactors with AGT
Serum-free, animal-origin-free, ultra-low protein formulation
VP SFM is formulated with an ultra-low protein concentration (< 5 μg/mL) for easier purification of recombinant proteins.
Easy adaptation for many popular cell lines
VP-SFM was designed for growth of VERO cells.  But it has also been used successfully to grow other cell lines (COS-7, MDCK, BHK-21, and Hep2) and for recombinant protein production, allowing the use of a single medium for multiple cell lines. VP-SFM is formulated without L-glutamine for greater stability and extended shelf life. Simply add L-glutamine or Glutamax supplement as needed.
cGMP manufacturing and quality system
VP-SFM is manufactured in a cGMP-compliant facility. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Scalability with Advanced Granulation Technology
Advanced Granulation Technology (AGT) helps simplify medium preparation at large scale with fast rehydration, a just-add-water protocol, and low dust generation, creating less pressure build-up during sterile filtration.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineMDCK, VERO, HEp2, BHK-21, COS-7
Endotoxin LevelLow
FormatSecuritainer Capped Vial
Included AntibioticsNo antibiotics
Product LineGibco™
Product TypeSerum Free Medium (SFM)
Quantity10 L
Reconstituted Volume10 L
Shipping ConditionWet Ice
ClassificationAnimal Origin-free, Chemically-defined, Protein-free, Serum-free
Culture TypeAdherent Cell Culture
FormPowder (AGT)
Serum LevelSerum-free
With AdditivesPhenol Red, Sodium Bicarbonate
Without AdditivesNo Glutamine
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.
Frequently asked questions (FAQs)
What are the main benefits of using AGT?
Complete formulations
- Homogeneous distribution of minute components
- Complete complex formulations in a dry format, including serum-free, protein-free, and chemically defined nutrient media
- Pre-adjusted to the appropriate pH and osmolality
- Require only standard supplementation with L-glutamine for L-glutamine dependent systems
- Can be customized for modified catalog and customers' proprietary formulations
Granular format
- Dissolves instantly for faster media preparation times
- Dust-free granules reduce mess and environmental contamination
- Performance equivalent to liquid media
- Workflow improvement
- Helps reduce total cycle costs due to lower number of components, decreasing time involved in raw material planning, procurement, and testing
- Faster and easier media preparation, with less potential for error
- Easy to scale up from research through production, in batch sizes from 2 kg to 6,000 kg
- Available in various packaging options
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is AGT?
Granulated cell culture media is an easy-to-use, granular form of dry media that is produced by applying fluid bed granulation technology to cell culture nutrient media manufacturing. The patented process allows production of complete formulations of a variety of serum-free, protein-free, and chemically defined media in a dry format.
AGT offers the benefits of liquid media without the cost, storage, and transportation issues. It is a complete medium that is pH and osmolality pre-adjusted. The granules dissolve instantly for faster media preparation time than conventional dry powder media. AGT media can therefore help reduce total cycle costs, decreasing time involved in raw material planning, procurement, and testing as well as media preparation. These benefits are all realized while maintaining comparable cell growth to liquid media.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link