Thermo Scientific Chemicals

2-Mesitylenesulfonyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: 125595000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

2-Mesitylenesulfonyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: 125595000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 125595000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
773-64-8
IUPAC Name
2,4,6-trimethylbenzene-1-sulfonyl chloride
Molecular Formula
C9H11ClO2S
InChI Key
PVJZBZSCGJAWNG-UHFFFAOYSA-N
SMILES
CC1=CC(C)=C(C(C)=C1)S(Cl)(=O)=O
Infrared spectrum
Conforms
Titration Argentometric
>=98.5 % (ex Cl)
Appearance (Form)
Crystalline powder
Melting point
53°C to 57°C
Appearance (Color)
White to grey

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text