Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
10 L
Packaging:
View all
Glass Bottle
Catalog number 125600025
Price (USD)/ Each
106.65
Online exclusive
125.00 
Save 18.35 (15%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
106.65
Online exclusive
125.00 
Save 18.35 (15%)
Add to cart
Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
Catalog number125600025
Price (USD)/ Each
106.65
Online exclusive
125.00 
Save 18.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name4-methylpent-3-en-2-one
Molecular FormulaC6H10O
InChI KeySHOJXDKTYKFBRD-UHFFFAOYSA-N
SMILESCC(C)=CC(C)=O
Molecular Weight (g/mol)98.15
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive index1.4420 to 1.4460 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only