Thermo Scientific Chemicals

Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals

Catalog number: 125600100
10 L, Plastic drum, Each
Thermo Scientific Chemicals

Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals

Catalog number: 125600100
10 L, Plastic drum, Each
Quantity
Packaging
Catalog number: 125600100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
141-79-7
IUPAC Name
4-methylpent-3-en-2-one
Molecular Formula
C6H10O
InChI Key
SHOJXDKTYKFBRD-UHFFFAOYSA-N
SMILES
CC(C)=CC(C)=O
Appearance (Color)
Clear colorless to yellow
Refractive index
1.4420 to 1.4460 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text