Methanesulfonic acid, 99%, extra pure, Thermo Scientific Chemicals
Methanesulfonic acid, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methanesulfonic acid, 99%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
5 mL
250 mL
1 L
Packaging:
View all
Plastic drum
Catalog number 125610051
Price (USD)/ Each
638.65
Online exclusive
709.00 
Save 70.35 (10%)
-
Add to cart
Quantity:
5 L
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
638.65
Online exclusive
709.00 
Save 70.35 (10%)
Add to cart
Methanesulfonic acid, 99%, extra pure, Thermo Scientific Chemicals
Catalog number125610051
Price (USD)/ Each
638.65
Online exclusive
709.00 
Save 70.35 (10%)
-
Add to cart
Chemical Identifiers
CAS75-75-2
IUPAC Namemethanesulfonic acid
Molecular FormulaCH4O3S
InChI KeyAFVFQIVMOAPDHO-UHFFFAOYSA-N
SMILESCS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<150 APHA
Titration with NaOH>=98.5 %
Infrared spectrumConforms
Appearance (Form)Clear liquid
Refractive index1.4285 to 1.4315 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only