Thermo Scientific Chemicals

Methanesulfonic acid, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 125612500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

Methanesulfonic acid, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 125612500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 125612500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-75-2
IUPAC Name
methanesulfonic acid
Molecular Formula
CH4O3S
InChI Key
AFVFQIVMOAPDHO-UHFFFAOYSA-N
SMILES
CS(O)(=O)=O
Color scale
=<150 APHA
Titration with NaOH
>=98.5 %
Infrared spectrum
Conforms
Appearance (Form)
Clear liquid
Refractive index
1.4285 to 1.4315 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text