TrypLE™ Select Enzyme (1X), no phenol red
TrypLE™ Select Enzyme (1X), no phenol red
Gibco™

TrypLE™ Select Enzyme (1X), no phenol red

Green features
Gibco™ TrypLE™ Select is an animal origin free, recombinant enzyme for dissociating a wide range of adherent mammalian cells.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
125630305 L
12563011100 mL
12563029500 mL
1256303110 L
Catalog number 12563030
Price (USD)
1,464.32
5 L
-
Add to cart
Quantity:
5 L
Price (USD)
1,464.32
5 L
Add to cart
Gibco TrypLE Select is an animal origin free, recombinant enzyme for dissociating a wide range of adherent mammalian cells, including CHO, HEK 293, A529, primary human keratinocytes, and embryonic stem cells. We offer a variety of TrypLE formulations for a range of cell culture applications.

Compared to trypsin and other dissociation reagents, TrypLE Select is:

  • Gentle on cells—maintains cell health for reproducible results
  • Room-temperature stable—convenient to use
  • Easy to use—substitutes directly into existing protocols with no inactivation required
  • Animal origin free (AOF)—pure, consistent, and effective

This TrypLE Select is modified as follows:

With

  • EDTA

Without

  • Phenol Red

Gentle on cells

TrypLE Select, like trypsin, cleaves peptide bonds on the C-terminal sides of lysine and arginine. However, TrypLE Select's exceptional purity increases specificity due to the action of a single enzyme. This reduces damage caused by cleavage from multiple enzymes in trypsin and other extracts.

Room-temperature stable

TrypLE Select is room-temperature stable and ready to use. TrypLE Select cell dissociation reagents remain stable for 24 months at room temperature or 2–8°C, making storage and handling easy and convenient.

Easy to use

TrypLE Select can be substituted for trypsin in existing protocols. In addition, dilution alone inactivates TrypLE Select, avoiding the need for trypsin inhibitors, such as FBS.

Animal Origin Free (AOF)

Unlike porcine trypsin and other enzymes, TrypLE Select is free of animal-derived components and is formulated on dedicated animal origin-free equipment.

Product Intended Use

Customers using TrypLE Select in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).

cGMP Manufacturing and Quality System

TrypLE Select is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

For Research Use or Further Manufacturing. Not for use in diagnostic procedures.
Specifications
Antibiotic SpectrumNo antibiotics
Cell LineVarious
Cell TypeMammalian Cells
Concentration1 X
Culture TypeAdherent
FormulationProprietary
Quantity5 L
Reagent TypeCell Dissociation
Serum LevelSerum-free
Shelf Life24 Months
ClassificationAnimal Origin-free
FormLiquid
Product TypeCell Culture Dissociation Reagent
SterilitySterile
Sterilization MethodSterile-filtered
With AdditivesNone
Without AdditivesNo Phenol Red
Unit Size5 L
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link