Thermo Scientific Chemicals

Methanesulfonyl chloride, 99.5%, Thermo Scientific Chemicals

Catalog number: 125640025
2.5 L, Glass Bottle, Each
Thermo Scientific Chemicals

Methanesulfonyl chloride, 99.5%, Thermo Scientific Chemicals

Catalog number: 125640025
2.5 L, Glass Bottle, Each
Quantity
Packaging
Catalog number: 125640025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
124-63-0
IUPAC Name
methanesulfonyl chloride
Molecular Formula
CH3ClO2S
InChI Key
QARBMVPHQWIHKH-UHFFFAOYSA-N
SMILES
CS(Cl)(=O)=O
Appearance (Form)
Clear liquid
Infrared spectrum
Conforms
Water
=<0.025 %
Color scale
=<100 APHA
GC
>=99.4 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text