1x1 image pixel for data collection
Methanesulfonyl chloride, 99.5%
Methanesulfonyl chloride, 99.5%
Methanesulfonyl chloride, 99.5%
Methanesulfonyl chloride, 99.5%
Thermo Scientific Chemicals

Methanesulfonyl chloride, 99.5%

CAS: 124-63-0 | CH3ClO2S | 114.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
250 mL
1 L
2.5 L
Packaging:
View all
Glass bottle
Catalog number 125641000
Price (USD)
35.65
Special offer
Online exclusive
Ends: 26-Sep-2025
43.80
Save 8.15 (19%)
Each
In stock
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
35.65
Special offer
Online exclusive
Ends: 26-Sep-2025
43.80
Save 8.15 (19%)
Each
Add to cart
Chemical Identifiers
CAS124-63-0
IUPAC Namemethanesulfonyl chloride
Molecular FormulaCH3ClO2S
InChI KeyQARBMVPHQWIHKH-UHFFFAOYSA-N
SMILESCS(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Color scale=<100 APHA
GC>=99.4 %
Refractive index1.4510 to 1.4530 (20°C, 589 nm)
Appearance (Form)Clear liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link