Thermo Scientific Chemicals

DL-Methionine, 99+%

Catalog number: 125652500
250 g, Plastic Bottle, Each
Thermo Scientific Chemicals

DL-Methionine, 99+%

Catalog number: 125652500
250 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 125652500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
59-51-8
IUPAC Name
2-amino-4-(methylsulfanyl)butanoic acid
Molecular Formula
C5H11NO2S
InChI Key
FFEARJCKVFRZRR-UHFFFAOYNA-N
SMILES
CSCCC(N)C(O)=O
Appearance (Form)
Crystalline powder or crystals or flakes
Titration with HClO4
99.0 to 101.0 % (on dried substance)
Sulfated ash
=<0.1 %
Chloride (Cl)
=<200 ppm
Related substances
=<0.2 % (TLC)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text