Thermo Scientific Chemicals

Methoxyacetyl chloride, 97%, stabilized, Thermo Scientific Chemicals

Catalog number: 125700100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Methoxyacetyl chloride, 97%, stabilized, Thermo Scientific Chemicals

Catalog number: 125700100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 125700100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
38870-89-2
IUPAC Name
2-methoxyacetyl chloride
Molecular Formula
C3H5ClO2
InChI Key
JJKWHOSQTYYFAE-UHFFFAOYSA-N
SMILES
COCC(Cl)=O
Infrared spectrum
Conforms
Refractive index
1.4190 to 1.4240 (20°C, 589 nm)
Stabilizer
0.3 % MgO
Titration RCOCl
>=96.0 %
Appearance (Color)
Clear colorless to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text