Thermo Scientific Chemicals

4-Methoxybenzamide, 97%, Thermo Scientific Chemicals

Catalog number: 125720250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Methoxybenzamide, 97%, Thermo Scientific Chemicals

Catalog number: 125720250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 125720250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3424-93-9
IUPAC Name
4-methoxybenzamide
Molecular Formula
C8H9NO2
InChI Key
GUCPYIYFQVTFSI-UHFFFAOYSA-N
SMILES
COC1=CC=C(C=C1)C(N)=O
Appearance (Color)
White
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
GC
>=96.0 %
Melting point
164°C to 167°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text