4-Methoxybenzenesulfonyl Chloride, 99%, Thermo Scientific Chemicals
4-Methoxybenzenesulfonyl Chloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methoxybenzenesulfonyl Chloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 125731000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS98-68-0
IUPAC Name4-methoxybenzene-1-sulfonyl chloride
Molecular FormulaC7H7ClO3S
InChI KeyDTJVECUKADWGMO-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline solid
GC>=98.5 %
Melting point36°C to 41°C
Appearance (Color)White to beige
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only