1x1 image pixel for data collection
SuperScript™ III One-Step RT-PCR System with Platinum™ Taq High Fidelity DNA Polymerase
SuperScript&trade; III One-Step RT-PCR System with Platinum&trade; <i>Taq</i> High Fidelity DNA Polymerase
Invitrogen™

SuperScript™ III One-Step RT-PCR System with Platinum™ Taq High Fidelity DNA Polymerase

The SuperScript III One-Step RT-PCR System with Platinum Taq High Fidelity is designed for convenient end-point detection and analysis ofRead more
Have Questions?
Catalog NumberNo. of Reactions
12574035100 Reactions
Catalog number 12574035
Price (USD)
1,097.65
Online Exclusive
1,246.00
Save 148.35 (12%)
Each
In stock
Add to cart
No. of Reactions:
100 Reactions
Request bulk or custom format
Price (USD)
1,097.65
Online Exclusive
1,246.00
Save 148.35 (12%)
Each
Add to cart

The SuperScript III One-Step RT-PCR System with Platinum Taq High Fidelity is designed for convenient end-point detection and analysis of RNA molecules by one-step RT-PCR. This one-step formulation allows for both cDNA synthesis and PCR amplification to be performed in a single tube using gene-specific primers and target RNAs from either total RNA or mRNA. It also allows for the detection of a wide range of RNA targets (up to 10 kb in length) at variable concentrations (1 pg to 1 μg of total RNA). This system consists of two major components: SuperScript III RT/Platinum Taq High Fidelity enzyme mix and 2X reaction mix.

SuperScript III RT is not significantly inhibited by ribosomal and transfer RNA and can be used to synthesize cDNA from total RNA. Platinum Taq DNA Polymerase High Fidelity is a an enzyme mixture composed of recombinant Taq DNA polymerase, Pyrococcus species GB–D polymerase, and Platinum Taq antibodies, which block polymerase activity at ambient temperatures enabling hot start PCR.

The 2X reaction mix consists of a proprietary buffer system optimized for reverse transcription and PCR amplification, Mg2+ optimized for universal use, deoxyribonucleotide triphosphates, and stabilizers.

The SuperScript III One-Step RT-PCR System with Platinum Taq High Fidelity offers:
• High-fidelity end-point detection and analysis of RNA molecules by RT-PCR
• PCR amplification and cDNA synthesis in a single tube
• Detection of a wide range of RNA targets from 300 bp to 10 kb
• Increased specificity with wide cDNA synthesis temperature range of 45−60°C

Note: For superior one-step RT-PCR performance, we recommend the SuperScript IV One-Step RT-PCR System or SuperScript IV One-Step RT-PCR System with ezDNase. The SuperScript IV One-Step RT-PCR System combines the high processivity of SuperScript IV Reverse Transcriptase with the high-fidelity of Platinum SuperFi DNA Polymerase to provide unmatched product yields, specificity, and sensitivity in less time and for a broad target range, even with suboptimally pure RNA samples.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypePCR Amplified cDNA
FormatOne-Step RT-PCR System
Hot StartBuilt-In Hot Start
No. of Reactions100 Reactions
Optimal Reaction Temperature50°C
PolymerasePlatinum Taq High Fidelity
Quantity100 rxns
Reaction FormatPremixed Components
Reagent TypeReverse Transcription
Reverse TranscriptaseSuperScript III
Ribonuclease H ActivityReduced
Shipping ConditionDry Ice
Size (Final Product)Up to 10 kb
Starting MaterialRNA
Technique1-Step RT-PCR
Detection MethodGel Electrophoresis
GC-Rich PCR PerformanceHigh
PCR Method1-step RT-PCR
Reaction Speed30 min.
Unit SizeEach
Contents & Storage

• SuperScript III RT/Platinum Taq High Fidelity Enzyme Mix (100 μL)
• 2X Reaction Mix (3 x 1 mL)
• 5 mM Magnesium Sulfate (500 μL)

Store at –10 to –30°C.

Frequently asked questions (FAQs)

How can I remove genomic DNA contamination from my sample prior to performing RT-PCR?

If amplification products are generated in the control tube/well that contains no reverse transcriptase (i.e., the no-RT control), it may be necessary to eliminate residual genomic DNA from the RNA sample. Use the following protocol to remove genomic DNA from the total RNA preparation.Random primers are the best choice for degraded RNA, RNA with heavy secondary structure, non-polyadenylated RNA, or prokaryotic RNA. It is recommended only for two-step RT-PCR, and typically gives the highest yields, although the cDNA may not necessarily be full length. Oligo(dT) primers are good to use when trying to recover full-length cDNA from 2-step RT-PCR. The reaction is influenced by secondary structure and RNA quality. Gene specific primers should be used for very specific, mainly one-step RT-PCR reactions. Random primers are the best choice for degraded RNA, RNA with heavy secondary structure, non-polyadenylated RNA, or prokaryotic RNA. It is recommended only for two-step RT-PCR, and typically gives the highest yields, although the cDNA may not necessarily be full length. Oligo(dT) primers are good to use when trying to recover full-length cDNA from 2-step RT-PCR. The reaction is influenced by secondary structure and RNA quality. Gene specific primers should be used for very specific, mainly one-step RT-PCR reactions.

Add the following to an autoclaved 0.5 mL microcentrifuge tube on ice:
1.Total RNA, ideally, less than or equal to 1 µg. (See Note 1 below.)
2.1.0 µL of 10X DNase buffer (200 mM Tris, pH 8.3, 500 mM KCl, 20 mM MgCl2).
3.0.1 U-3.0 U of DNase I (RNase-free, Cat. No. 18047019) or 1.0 U Dnase I, Amplification Grade (Cat. No. 18068015. (See Note 2 below.)
4.Bring volume up to 10 µL with DEPC-treated water.
5.Incubate at room temperature for 15 min. (See Note 3 below.)
6.Terminate the reaction by adding 1 µL 25 mM EDTA and heat 10 min at 65 degrees C. (See Note 4 below.)
7.Place on ice for 1 minute.
8.Collect by brief centrifugation. This mixture can be used directly for reverse transcription.

Please note the following:
1.To work with higher quantities of RNA, scale up the entire reaction linearly. Do not exceed 2 µg RNA in the 10 µL reaction. More RNA will increase the viscosity of the solution and prevent the DNAse I from diffusing and finding the DNA.
2.DNAse I, Amplification Grade has been extensively purified to remove trace ribonuclease activities commonly associated with other "RNAse-free" enzyme preparations and does not require the addition of placental RNAse inhibitor.
3.It is important not to exceed the 15 minute incubation time or the room temperature incubation. Higher temperatures and longer times could lead to Mg2+-dependent hydrolysis of the RNA.
4.This procedure requires careful pipetting of all solutions so that the concentration of divalent metal cation (Mg2+) is controlled.
5.Because the DNAse I must be heated to 65 degrees C to inactivate the enzyme, the concentration of free divalent metal ions must be low enough (less than 1 mM) after addition of the EDTA to prevent chemical hydrolysis of the RNA. See references below.
After the addition of EDTA, there is an approximately 1:1 molar ratio of Mg2+ :EDTA. EDTA chelates Mg2+ molecules on a 1:1 molar basis. Therefore, this RNA can be directly used in a reverse transcription reaction. First-strand reverse transcription buffers typically result in a final concentration of 2.5 mM Mg2+. If the reverse transcription buffer does not contain MgCl2, add it to the reaction at a final concentration of 2.5 mM. This results in a net final concentration of approximately 2.25 to 2.5 mM MgCl2.

References on RNA hydrolysis:
Molekulyarnaya Biologiya (1987) 21:1235-1241.
References on the mechanism of hydrolysis by other cations:
Eichorn GL and Butzov JY (1965) Biopolymers 3:79.
Butzov JY and Eichorn GL (1965) Biopolymers 3:95.
Farkas WR (1968) Biochim Biophys Acta 155:401.
The authors of the first paper express the opinion that the mechanism of the nonspecific hydrolysis by cations which proceeds through 2',3' cyclic phosphate formation is similar to that of specific hydrolysis such as RNA splicing.

How much RNA should be employed for first-strand cDNA synthesis?

The amount of RNA template for a cDNA synthesis is highly flexible and depends upon the amount of sample available and an individual's need. In general, 1 µg total RNA is used in a typical 20-µL RT reaction.

Find additional tips, troubleshooting help, and resources within ourReverse Transcription and RACE Support Center.

Should I treat the cDNA with RNase H prior to downstream processing?

Some feel that the RNA in the RNA:DNA duplex after reverse transcription will inhibit PCR primers from annealing and amplifying the cDNA. The RNA is still present when using RNase H-mutant RTs. RNase H frees the cDNA from the RNA. On the other hand, some feel that the 95 degrees C denaturing step will cause the RNA primers to fall off the DNA and therefore RNase H treatment is not necessary. Therefore, this step is optional. For cloning of larger fragments, RNase H treatment can be beneficial.

What percentage of RNA is converted to cDNA when performing reverse transcription?

This depends highly on the quality of the sample. mRNA itself makes up 1-5% of total RNA. Depending on the primer and enzyme used, reverse transcription can covert >70% of that into cDNA.

Find additional tips, troubleshooting help, and resources within our Reverse Transcription and RACE Support Center.

I'm setting up my RT reaction and am trying to decide whether I should use random primers, oligo(dT) primer, gene-specific primer, or oligo(dT)/random mix primers. What would you suggest?

Random primers are the best choice for degraded RNA, RNA with heavy secondary structure, non-polyadenylated RNA, or prokaryotic RNA. It is recommended only for two-step RT-PCR, and typically gives the highest yields, although the cDNA may not necessarily be full length. Oligo(dT) primers are good to use when trying to recover full-length cDNA from 2-step RT-PCR. The reaction is influenced by secondary structure and RNA quality. Gene specific primers should be used for very specific, mainly one-step RT-PCR reactions.

Find additional tips, troubleshooting help, and resources within our Reverse Transcription and RACE Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection