2-Methoxybenzyl alcohol, 99%, Thermo Scientific Chemicals
2-Methoxybenzyl alcohol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methoxybenzyl alcohol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Catalog number 125760100
Price (USD)/ Each
-
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS612-16-8
IUPAC Name(2-methoxyphenyl)methanol
Molecular FormulaC8H10O2
InChI KeyWYLYBQSHRJMURN-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to Yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5460 to 1.5490 (20°C, 589 nm)
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only