4-Methoxybenzylamine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Methoxybenzylamine, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
500 mL
Packaging:
View all
Glass bottle
Catalog number 125781000
Price (USD)/ Each
123.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
145.00 
Save 21.35 (15%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
123.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
145.00 
Save 21.35 (15%)
Add to cart
4-Methoxybenzylamine, 98%, Thermo Scientific Chemicals
Catalog number125781000
Price (USD)/ Each
123.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
145.00 
Save 21.35 (15%)
-
Add to cart
Chemical Identifiers
CAS2393-23-9
IUPAC Name1-(4-methoxyphenyl)methanamine
Molecular FormulaC8H11NO
InChI KeyIDPURXSQCKYKIJ-UHFFFAOYSA-N
SMILESCOC1=CC=C(CN)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
GC>=97.5 %
Infrared spectrumConforms
Refractive index1.5460 to 1.5480 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only