p-Methoxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
p-Methoxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals

p-Methoxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 125841000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name(2E)-3-(4-methoxyphenyl)prop-2-enoic acid
Molecular FormulaC10H10O3
InChI KeyAFDXODALSZRGIH-QPJJXVBHSA-N
SMILESCOC1=CC=C(\C=C\C(O)=O)C=C1
Molecular Weight (g/mol)178.19
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to light brown
Appearance (Form)Powder, crystals and/or chunks
Infrared spectrumConforms
Melting point171°C to 176°C
Titration with NaOH97.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only