Bis(2-methoxyethyl) ether, 99%, extra pure, Thermo Scientific Chemicals
Bis(2-methoxyethyl) ether, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(2-methoxyethyl) ether, 99%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
2.5 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 125850010
Price (USD)/ Each
112.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
132.00 
Save 19.35 (15%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
112.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
132.00 
Save 19.35 (15%)
Add to cart
Bis(2-methoxyethyl) ether, 99%, extra pure, Thermo Scientific Chemicals
Catalog number125850010
Price (USD)/ Each
112.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
132.00 
Save 19.35 (15%)
-
Add to cart
Chemical Identifiers
CAS111-96-6
IUPAC Name1-methoxy-2-(2-methoxyethoxy)ethane
Molecular FormulaC6H14O3
InChI KeySBZXBUIDTXKZTM-UHFFFAOYSA-N
SMILESCOCCOCCOC
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.025 %
Appearance (Form)Clear liquid
Infrared spectrumConforms
GC>=98.5 %
Color scale=<10 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only