Thermo Scientific Chemicals

3-Methoxyphenylacetic acid, 99.5%, Thermo Scientific Chemicals

Catalog number: 126010250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Methoxyphenylacetic acid, 99.5%, Thermo Scientific Chemicals

Catalog number: 126010250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 126010250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1798-09-0
IUPAC Name
2-(3-methoxyphenyl)acetate
Molecular Formula
C9H9O3
InChI Key
LEGPZHPSIPPYIO-UHFFFAOYSA-M
SMILES
COC1=CC=CC(CC([O-])=O)=C1
Melting point
68°C to 72°C
Titration with NaOH
>=99.4 %
Appearance (Color)
White to light beige
Appearance (Form)
Powder and/or chunks
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text