3-Methoxyphenylacetic acid, 99.5%, Thermo Scientific Chemicals
3-Methoxyphenylacetic acid, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methoxyphenylacetic acid, 99.5%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 126011000
Price (USD)/ Each
283.00
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
283.00
Add to cart
3-Methoxyphenylacetic acid, 99.5%, Thermo Scientific Chemicals
Catalog number126011000
Price (USD)/ Each
283.00
-
Add to cart
Chemical Identifiers
IUPAC Name2-(3-methoxyphenyl)acetate
Molecular FormulaC9H9O3
InChI KeyLEGPZHPSIPPYIO-UHFFFAOYSA-M
SMILESCOC1=CC=CC(CC([O-])=O)=C1
Molecular Weight (g/mol)165.17
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point68°C to 72°C
Titration with NaOH>=99.4 %
Appearance (Color)White to light beige
Appearance (Form)Powder and/or chunks
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only