Sample and Skimmer Cones
Sample and Skimmer Cones
Actual product may vary
Thermo Scientific™
Sample and Skimmer Cones
Choose from this range of sample and skimmer cones to suit your analysis requirements when using the Thermo Scientific™ ELEMENT™ Series of HR-ICP-MS instruments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeMaterial
1260630Jet Sample ConeNickel
1044530Sample ConeNickel
1047460Skimmer ConePlatinum, Boron-free
1047461Skimmer ConePlatinum
1067500Sample ConePlatinum, Boron-free
1067501Sample ConePlatinum
1067600Skimmer ConeNickel
1142150Skimmer ConePlatinum, X-Vers, Boron-free
1142151Skimmer ConePlatinum, X-Version
1142160Skimmer ConeNickel, X-Version
1184050Sample ConeAluminum
1184060Skimmer ConeAluminum
BRE0010791Jet Sample ConePlatinum, Boron-free
BRE0010792Jet Sample ConePlatinum w/ Boron
Catalog number 1260630
Price (USD)
-
Request A Quote
Type:
Jet Sample Cone
Material:
Nickel
Ask our AI about this Product
A range of easily changeable Ni, Pt, and Al Thermo Scientific™ Sample and Skimmer Cones are available for the Thermo Scientific™ ELEMENT™ Series HR-ICP-MS and Thermo Scientific™ Neptune™ Series MC-ICP-MS systems to suit your analysis requirements, all designed for durability, performance and ease of use.
Select High-Quality Consumables for Consistent Performance
These high-quality, field-proven cones are designed for reliable high-sensitivity analysis down to detection-limit level by ICP-MS. Available in solid Ni construction, Pt-tipped for even greater durability, and Al for specific analysis requirements.
Specifications
For Use With (Equipment)ELEMENT Series HR-ICP-MS System and Neptune Series MC-ICP-MS System
MaterialNickel
TypeJet Sample Cone
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link