Thermo Scientific Chemicals

4'-Methoxypropiophenone, 99%, Thermo Scientific Chemicals

Catalog number: 126091000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

4'-Methoxypropiophenone, 99%, Thermo Scientific Chemicals

Catalog number: 126091000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 126091000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
121-97-1
IUPAC Name
1-(4-methoxyphenyl)propan-1-one
Molecular Formula
C10H12O2
InChI Key
ZJVAWPKTWVFKHG-UHFFFAOYSA-N
SMILES
CCC(=O)C1=CC=C(OC)C=C1
Infrared spectrum
Conforms
Refractive index
1.5450 to 1.5480 (20°C, 589 nm)
GC
>=98.5 %
Appearance (Color)
Colorless to white to yellow to amber
Appearance (Form)
Clear liquid or low melting solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text