Thermo Scientific Chemicals

Methyl acetoacetate, 99+%, Thermo Scientific Chemicals

Catalog number: 126150010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

Methyl acetoacetate, 99+%, Thermo Scientific Chemicals

Catalog number: 126150010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 126150010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
105-45-3
IUPAC Name
methyl 3-oxobutanoate
Molecular Formula
C5H8O3
InChI Key
WRQNANDWMGAFTP-UHFFFAOYSA-N
SMILES
COC(=O)CC(C)=O
Appearance (Form)
Clear liquid
Refractive index
1.4175 to 1.4200 (20°C, 589 nm)
Free acid (CH3COOH)
=<0.2 %
Color scale
=<20 APHA
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text